The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080657.4 | NP_542388.2 | 3 - 4 | 876 | 104 |
XM_011510414.2 | XP_011508716.1 | 4 - 5 | 1965 | 104 | |
XM_011510415.2 | XP_011508717.1 | 3 - 4 | 707 | 104 | |
GenBank mRNA | AF442151.1 | - | 3 - 4 | 764 | 104 |
AK300085.1 | - | 3 - 4 | 642 | 104 | |
AK312670.1 | - | 3 - 4 | 775 | 104 | |
BC017969.1 | - | 3 - 4 | 764 | 104 |