The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040118.2 | NP_001035207.1 | 22 - 23 | 3355 | 70 |
NM_001135190.1 | NP_001128662.1 | 19 - 20 | 2983 | 70 | |
NM_015242.4 | NP_056057.2 | 20 - 21 | 3166 | 70 | |
GenBank mRNA | AB018325.2 | - | 20 - 21 | 3143 | 70 |
AF411983.1 | - | 16 - 17 | 2224 | 70 | |
AJ621557.1 | - | 20 - 21 | 3166 | 70 | |
AY049732.1 | - | 17 - 18 | 3003 | 70 | |
AY553630.1 | - | 22 - 23 | 3370 | 70 | |
BC008315.1 | - | 4 - 5 | 636 | 70 | |
BC056401.1 | - | 8 - 9 | 1009 | 70 | |
BC140792.1 | - | 19 - 20 | 2653 | 70 | |
BC144562.1 | - | 20 - 21 | 2836 | 70 |