Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARAP1
Entrez Gene ID
116985
Gene Name
ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1
Gene Aliases
CENTD2
Chromosome Location
Chr.11: 72685069 - 72752389 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040118.2 NP_001035207.1 7 - 8 1215 63
NM_001135190.1 NP_001128662.1 5 - 6 1026 63
NM_015242.4 NP_056057.2 5 - 6 1026 63
GenBank mRNA AB018325.2 - 5 - 6 1003 63
AF411983.1 - 1 - 2 84 63
AJ621557.1 - 5 - 6 1026 63
AK160383.1 - 4 - 5 1364 63
AY049732.1 - 2 - 3 863 63
AY553630.1 - 7 - 8 1230 63
BC140792.1 - 5 - 6 696 63
BC144562.1 - 5 - 6 696 63

View Full Product Details