Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDCD1LG2
Entrez Gene ID
80380
Gene Name
programmed cell death 1 ligand 2
Gene Aliases
B7DC, Btdc, CD273, PD-L2, PDCD1L2, PDL2, bA574F11.2
Chromosome Location
Chr.9: 5510438 - 5571282 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025239.3 NP_079515.2 3 - 4 629 82
XM_005251600.3 XP_005251657.1 3 - 4 736 82
GenBank mRNA AF329193.1 - 3 - 4 614 82
AF344424.1 - 3 - 4 629 82
AK001872.1 - 1 - 2 60 82
AY254343.1 - 2 - 3 356 82
BC074766.2 - 3 - 4 390 82
BC113678.1 - 3 - 4 392 82
BC113680.1 - 3 - 4 392 82
BC143920.1 - 3 - 4 392 82
BX951459.1 - 1 - 2 266 82
DQ336699.1 - 2 - 3 323 82
EF444806.1 - 3 - 4 476 82

View Full Product Details