Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MANSC4
Entrez Gene ID
100287284
Gene Name
MANSC domain containing 4
Gene Aliases
-
Chromosome Location
Chr.12: 27762645 - 27771899 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146221.1 NP_001139693.1 2 - 3 361 73
XM_011520542.2 XP_011518844.1 3 - 4 907 73

View Full Product Details