The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020124.2 | NP_064509.2 | 1 - 1 | 471 | 120 |
GenBank mRNA | AF146759.1 | - | 1 - 1 | 493 | 120 |
AF315688.1 | - | 1 - 1 | 482 | 120 | |
AY358855.1 | - | 1 - 1 | 449 | 120 |