Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- MARK3
- Entrez Gene ID
-
4140
- Gene Name
- microtubule affinity regulating kinase 3
- Gene Aliases
-
CTAK1, KP78, PAR1A, Par-1a
- Chromosome Location
-
Chr.14: 103385364 - 103503831 on Build GRCh38
View Full Product Details