The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000833.4 | NP_000824.1 | 1 - 2 | 107 | 139 |
XM_011522461.2 | XP_011520763.1 | 1 - 2 | 717 | 139 | |
GenBank mRNA | U90277.1 | - | 1 - 2 | 107 | 139 |