The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297654.1 | NP_001284583.1 | 11 - 12 | 1729 | 53 |
NM_013993.2 | NP_054699.2 | 12 - 13 | 1773 | 53 | |
NM_013994.2 | NP_054700.2 | 10 - 11 | 1556 | 53 | |
XM_006715185.2 | XP_006715248.1 | 11 - 12 | 1720 | 53 | |
XM_011514882.2 | XP_011513184.1 | 11 - 12 | 1720 | 53 | |
XM_011514883.1 | XP_011513185.1 | 13 - 14 | 1835 | 53 | |
XM_011514884.1 | XP_011513186.1 | 13 - 14 | 1911 | 53 | |
XM_011514885.1 | XP_011513187.1 | 11 - 12 | 1758 | 53 | |
XM_011514886.1 | XP_011513188.1 | 11 - 12 | 1623 | 53 | |
XM_011514887.1 | XP_011513189.1 | 11 - 12 | 1773 | 53 | |
XM_017011268.1 | XP_016866757.1 | 11 - 12 | 1590 | 53 | |
GenBank mRNA | AK130776.1 | - | 6 - 7 | 820 | 53 |
L11315.1 | - | 12 - 13 | 1655 | 53 | |
L20817.1 | - | 11 - 12 | 1706 | 53 |