The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001202521.1 | NP_001189450.1 | 13 - 14 | 2133 | 128 |
NM_001202522.1 | NP_001189451.1 | 13 - 14 | 2051 | 128 | |
NM_001202523.1 | NP_001189452.1 | 15 - 16 | 2630 | 128 | |
NM_001297652.1 | NP_001284581.1 | 15 - 16 | 2749 | 128 | |
NM_001297653.1 | NP_001284582.1 | 15 - 16 | 2551 | 128 | |
NM_001297654.1 | NP_001284583.1 | 16 - 17 | 2662 | 128 | |
NM_001954.4 | NP_001945.3 | 17 - 18 | 2669 | 128 | |
NM_013993.2 | NP_054699.2 | 17 - 18 | 2706 | 128 | |
NM_013994.2 | NP_054700.2 | 15 - 16 | 2507 | 128 | |
XM_006715185.2 | XP_006715248.1 | 16 - 17 | 2653 | 128 | |
XM_011514882.2 | XP_011513184.1 | 16 - 17 | 2671 | 128 | |
XM_011514883.1 | XP_011513185.1 | 18 - 19 | 2786 | 128 | |
XM_011514884.1 | XP_011513186.1 | 18 - 19 | 2862 | 128 | |
XM_011514885.1 | XP_011513187.1 | 16 - 17 | 2709 | 128 | |
XM_011514886.1 | XP_011513188.1 | 16 - 17 | 2574 | 128 | |
XM_011514887.1 | XP_011513189.1 | 16 - 17 | 2724 | 128 | |
XM_011514888.2 | XP_011513190.1 | 15 - 16 | 2560 | 128 | |
XM_017011268.1 | XP_016866757.1 | 16 - 17 | 2541 | 128 | |
XM_017011269.1 | XP_016866758.1 | 15 - 16 | 2613 | 128 | |
GenBank mRNA | AB210021.1 | - | 16 - 17 | 2490 | 128 |
AK291621.1 | - | 15 - 16 | 2444 | 128 | |
AK294793.1 | - | 15 - 16 | 2616 | 128 | |
BC008716.2 | - | 17 - 18 | 2669 | 128 | |
BC013400.2 | - | 17 - 18 | 2669 | 128 | |
BC070070.1 | - | 15 - 16 | 2440 | 128 | |
BI193181.1 | - | 3 - 4 | 549 | 127 | |
L11315.1 | - | 17 - 18 | 2588 | 128 | |
L20817.1 | - | 16 - 17 | 2639 | 128 | |
L57508.1 | - | 15 - 16 | 2528 | 128 | |
X74979.1 | - | 14 - 15 | 2381 | 128 | |
Z29093.1 | - | 14 - 15 | 2672 | 128 |