Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDR1
Entrez Gene ID
780
Gene Name
discoidin domain receptor tyrosine kinase 1
Gene Aliases
CAK, CD167, DDR, EDDR1, HGK2, MCK10, NEP, NTRK4, PTK3, PTK3A, RTK6, TRKE
Chromosome Location
Chr.6: 30880909 - 30900156 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202521.1 NP_001189450.1 3 - 4 463 90
NM_001202522.1 NP_001189451.1 3 - 4 463 90
NM_001202523.1 NP_001189452.1 4 - 5 715 90
NM_001297652.1 NP_001284581.1 4 - 5 834 90
NM_001297653.1 NP_001284582.1 4 - 5 636 90
NM_001297654.1 NP_001284583.1 4 - 5 636 90
NM_001954.4 NP_001945.3 6 - 7 754 90
NM_013993.2 NP_054699.2 5 - 6 680 90
NM_013994.2 NP_054700.2 3 - 4 463 90
XM_006715185.2 XP_006715248.1 4 - 5 627 90
XM_011514882.2 XP_011513184.1 4 - 5 627 90
XM_011514883.1 XP_011513185.1 6 - 7 742 90
XM_011514884.1 XP_011513186.1 6 - 7 818 90
XM_011514885.1 XP_011513187.1 4 - 5 665 90
XM_011514886.1 XP_011513188.1 4 - 5 530 90
XM_011514887.1 XP_011513189.1 4 - 5 680 90
XM_011514888.2 XP_011513190.1 4 - 5 627 90
XM_017011268.1 XP_016866757.1 4 - 5 497 90
XM_017011269.1 XP_016866758.1 4 - 5 680 90
GenBank mRNA AB210021.1 - 5 - 6 575 90
AK291621.1 - 4 - 5 529 90
AK294793.1 - 4 - 5 701 90
AK295643.1 - 4 - 5 528 90
BC008716.2 - 6 - 7 754 90
BC013400.2 - 6 - 7 754 90
BC070070.1 - 4 - 5 525 90
EU826613.1 - 3 - 4 421 90
EU826614.1 - 3 - 4 421 90
L11315.1 - 5 - 6 562 90
L20817.1 - 4 - 5 613 90
L57508.1 - 4 - 5 613 90
X74979.1 - 3 - 4 466 90
Z29093.1 - 3 - 4 757 90

View Full Product Details