The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242832.1 | NP_001229761.1 | 2 - 3 | 463 | 68 |
NM_032389.4 | NP_115765.2 | 2 - 3 | 463 | 68 | |
XM_005253166.1 | XP_005253223.1 | 2 - 3 | 841 | 68 | |
XM_005253167.1 | XP_005253224.1 | 2 - 3 | 841 | 68 | |
XM_005253168.2 | XP_005253225.1 | 2 - 3 | 841 | 68 | |
XM_006718346.1 | XP_006718409.1 | 2 - 3 | 841 | 68 | |
XM_011520405.1 | XP_011518707.1 | 2 - 3 | 841 | 68 | |
XM_017018413.1 | XP_016873902.1 | 2 - 3 | 841 | 68 | |
XM_017018414.1 | XP_016873903.1 | 2 - 3 | 217 | 68 | |
XM_017018415.1 | XP_016873904.1 | 2 - 3 | 211 | 68 | |
GenBank mRNA | AB073358.1 | - | 1 - 2 | 73 | 68 |
AK027482.1 | - | 2 - 3 | 199 | 68 | |
AK098324.1 | - | 2 - 3 | 217 | 68 | |
AK300347.1 | - | 2 - 3 | 197 | 68 | |
AK301785.1 | - | 2 - 3 | 167 | 68 | |
AK304541.1 | - | 2 - 3 | 202 | 68 | |
AL834337.1 | - | 2 - 3 | 182 | 68 | |
BC030148.2 | - | 2 - 3 | 206 | 68 | |
BC036823.1 | - | 2 - 3 | 208 | 68 |