Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TRAPPC10
- Entrez Gene ID
-
7109
- Gene Name
- trafficking protein particle complex 10
- Gene Aliases
-
EHOC-1, EHOC1, GT334, TMEM1, TRS130, TRS30
- Chromosome Location
-
Chr.21: 44012325 - 44106552 on Build GRCh38
View Full Product Details