Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAPPC10
Entrez Gene ID
7109
Gene Name
trafficking protein particle complex 10
Gene Aliases
EHOC-1, EHOC1, GT334, TMEM1, TRS130, TRS30
Chromosome Location
Chr.21: 44012325 - 44106552 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003274.4 NP_003265.3 9 - 10 1470 123
XM_005261168.4 XP_005261225.1 9 - 10 1463 123
XM_011529714.2 XP_011528016.1 9 - 10 1463 123
XM_011529715.2 XP_011528017.1 9 - 10 1463 123
XM_011529716.2 XP_011528018.1 8 - 9 1316 123
XM_011529717.2 XP_011528019.1 9 - 10 1463 123
XM_011529718.2 XP_011528020.1 9 - 10 1463 123
XM_011529719.2 XP_011528021.1 7 - 8 1019 123
XM_011529720.2 XP_011528022.1 9 - 10 1463 123
XM_011529721.2 XP_011528023.1 6 - 7 836 123
XM_011529722.2 XP_011528024.1 6 - 7 802 123
XM_011529723.2 XP_011528025.1 5 - 6 699 123
XM_011529725.2 XP_011528027.1 9 - 10 1463 123
XM_011529726.2 XP_011528028.1 9 - 10 1463 123
XM_011529727.2 XP_011528029.1 5 - 6 704 123
XM_011529728.2 XP_011528030.1 7 - 8 1096 123
XM_011529729.2 XP_011528031.1 5 - 6 609 123
XM_011529730.2 XP_011528032.1 9 - 10 1463 123
XM_017028454.1 XP_016883943.1 8 - 9 1316 123
GenBank mRNA BC101728.1 - 9 - 10 1390 123
U19252.1 - 9 - 10 1438 123
U61500.1 - 9 - 10 1454 123

View Full Product Details