Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ROBO4
Entrez Gene ID
54538
Gene Name
roundabout guidance receptor 4
Gene Aliases
ECSM4, MRB
Chromosome Location
Chr.11: 124884218 - 124897935 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301088.1 NP_001288017.1 14 - 15 2133 95
NM_019055.5 NP_061928.4 14 - 15 2339 95
XM_011542875.1 XP_011541177.1 8 - 9 1101 95
GenBank mRNA AF361473.1 - 14 - 15 2268 95
AK000805.1 - 6 - 7 795 95
AK025195.1 - 2 - 3 189 95
AK027852.1 - 14 - 15 2237 95
AK074163.1 - 13 - 14 1869 95
AK289769.1 - 14 - 15 2238 95
AK302628.1 - 14 - 15 2003 95
AY358083.1 - 14 - 15 2190 95
BC014995.1 - 13 - 14 1951 95
BC039602.1 - 14 - 15 2227 95
BC064643.1 - 4 - 5 486 95
BC111562.1 - 14 - 15 2233 95
BC111748.1 - 14 - 15 2233 95

View Full Product Details