Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ROBO4
Entrez Gene ID
54538
Gene Name
roundabout guidance receptor 4
Gene Aliases
ECSM4, MRB
Chromosome Location
Chr.11: 124884218 - 124897935 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301088.1 NP_001288017.1 7 - 8 1079 70
NM_019055.5 NP_061928.4 7 - 8 1285 70
XM_006718861.1 XP_006718924.1 7 - 8 1218 70
XM_011542875.1 XP_011541177.1 1 - 2 52 70
GenBank mRNA AF361473.1 - 7 - 8 1214 70
AK027852.1 - 7 - 8 1184 70
AK074163.1 - 6 - 7 815 70
AK289769.1 - 7 - 8 1184 70
AK297845.1 - 1 - 2 52 70
AK302628.1 - 7 - 8 949 70
AY358083.1 - 7 - 8 1136 70
BC014995.1 - 6 - 7 897 70
BC039602.1 - 7 - 8 1173 70
BC111562.1 - 7 - 8 1179 70
BC111748.1 - 7 - 8 1179 70

View Full Product Details