The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005502.3 | NP_005493.2 | 11 - 12 | 1716 | 76 |
XM_005251773.2 | XP_005251830.1 | 11 - 12 | 1625 | 76 | |
XM_005251776.2 | XP_005251833.1 | 10 - 11 | 1465 | 76 | |
XM_011518339.2 | XP_011516641.1 | 12 - 13 | 1700 | 76 | |
XM_011518340.2 | XP_011516642.1 | 13 - 14 | 1915 | 76 | |
XM_011518341.2 | XP_011516643.1 | 12 - 13 | 1700 | 76 | |
XM_011518342.2 | XP_011516644.1 | 9 - 10 | 1403 | 76 | |
XM_011518344.2 | XP_011516646.1 | 12 - 13 | 1700 | 76 | |
XM_017014378.1 | XP_016869867.1 | 12 - 13 | 1898 | 76 | |
XM_017014379.1 | XP_016869868.1 | 12 - 13 | 2306 | 76 | |
XM_017014380.1 | XP_016869869.1 | 12 - 13 | 1632 | 76 | |
XM_017014381.1 | XP_016869870.1 | 12 - 13 | 1698 | 76 | |
XM_017014382.1 | XP_016869871.1 | 10 - 11 | 2163 | 76 | |
GenBank mRNA | AB055982.1 | - | 10 - 11 | 1312 | 76 |
AB445477.1 | - | 10 - 11 | 1312 | 76 | |
AF165281.1 | - | 9 - 10 | 1252 | 76 | |
AF285167.1 | - | 11 - 12 | 1602 | 76 | |
AJ012376.1 | - | 9 - 10 | 1252 | 76 | |
BC146856.1 | - | 11 - 12 | 1526 | 76 |