The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005502.3 | NP_005493.2 | 15 - 16 | 2521 | 63 |
XM_005251773.2 | XP_005251830.1 | 15 - 16 | 2430 | 63 | |
XM_005251776.2 | XP_005251833.1 | 14 - 15 | 2270 | 63 | |
XM_011518339.2 | XP_011516641.1 | 16 - 17 | 2505 | 63 | |
XM_011518340.2 | XP_011516642.1 | 17 - 18 | 2720 | 63 | |
XM_011518341.2 | XP_011516643.1 | 16 - 17 | 2505 | 63 | |
XM_011518342.2 | XP_011516644.1 | 13 - 14 | 2208 | 63 | |
XM_011518344.2 | XP_011516646.1 | 16 - 17 | 2505 | 63 | |
XM_017014378.1 | XP_016869867.1 | 16 - 17 | 2703 | 63 | |
XM_017014379.1 | XP_016869868.1 | 16 - 17 | 3111 | 63 | |
XM_017014380.1 | XP_016869869.1 | 16 - 17 | 2437 | 63 | |
XM_017014381.1 | XP_016869870.1 | 16 - 17 | 2503 | 63 | |
XM_017014382.1 | XP_016869871.1 | 14 - 15 | 2968 | 63 | |
GenBank mRNA | AB055982.1 | - | 14 - 15 | 2117 | 63 |
AB445477.1 | - | 14 - 15 | 2117 | 63 | |
AF165281.1 | - | 13 - 14 | 2057 | 63 | |
AF285167.1 | - | 15 - 16 | 2407 | 63 | |
AJ012376.1 | - | 13 - 14 | 2057 | 63 | |
BC146856.1 | - | 15 - 16 | 2331 | 63 |