Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC19A3
Entrez Gene ID
80704
Gene Name
solute carrier family 19 member 3
Gene Aliases
BBGD, THMD2, THTR2
Chromosome Location
Chr.2: 227685210 - 227718030 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025243.3 NP_079519.1 3 - 4 1072 69
XM_005246874.3 XP_005246931.1 4 - 5 1235 69
XM_011511931.2 XP_011510233.1 3 - 4 1327 69
XM_011511932.1 XP_011510234.1 3 - 4 1149 69
XM_011511933.1 XP_011510235.1 3 - 4 1415 69
XM_017005030.1 XP_016860519.1 4 - 5 1417 69
XM_017005031.1 XP_016860520.1 4 - 5 1396 69
XM_017005032.1 XP_016860521.1 4 - 5 1277 69
XM_017005033.1 XP_016860522.1 4 - 5 1591 69
XM_017005034.1 XP_016860523.1 4 - 5 1521 69
GenBank mRNA AF271633.1 - 3 - 4 1026 69
AF283317.1 - 3 - 4 1053 69
AK301490.1 - 4 - 5 1238 69
AK312464.1 - 3 - 4 1074 69
BC032014.1 - 3 - 4 1027 69

View Full Product Details