The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166356.1 | NP_001159828.1 | 6 - 7 | 898 | 76 |
NM_001166357.1 | NP_001159829.1 | 6 - 7 | 990 | 76 | |
NM_001166358.1 | NP_001159830.1 | 6 - 7 | 889 | 76 | |
NM_001166359.1 | NP_001159831.1 | 6 - 7 | 782 | 76 | |
NM_005412.5 | NP_005403.2 | 6 - 7 | 928 | 76 | |
NR_029415.1 | 5 - 6 | 730 | 76 | ||
NR_029416.1 | 5 - 6 | 809 | 76 | ||
NR_029417.1 | 5 - 6 | 848 | 76 | ||
NR_048562.1 | 5 - 6 | 938 | 76 | ||
XM_011538675.2 | XP_011536977.1 | 6 - 7 | 868 | 76 | |
XM_011538676.2 | XP_011536978.1 | 6 - 7 | 859 | 76 | |
XM_011538677.2 | XP_011536979.1 | 5 - 6 | 769 | 76 | |
XM_011538678.2 | XP_011536980.1 | 5 - 6 | 751 | 76 | |
GenBank mRNA | AK055053.1 | - | 3 - 4 | 1578 | 76 |
AK223555.1 | - | 6 - 7 | 790 | 76 | |
AK295941.1 | - | 5 - 6 | 679 | 76 | |
AK296183.1 | - | 6 - 7 | 742 | 76 | |
AK297173.1 | - | 5 - 6 | 690 | 76 | |
AK298231.1 | - | 5 - 6 | 583 | 76 | |
AK301339.1 | - | 6 - 7 | 655 | 76 | |
AK301443.1 | - | 5 - 6 | 752 | 76 | |
AK303827.1 | - | 5 - 6 | 604 | 76 | |
AK315916.1 | - | 6 - 7 | 982 | 76 | |
BC011911.2 | - | 6 - 7 | 732 | 76 | |
BC013677.2 | - | 6 - 7 | 792 | 76 | |
BC025355.1 | - | 3 - 4 | 315 | 76 | |
BC032584.1 | - | 6 - 7 | 771 | 76 | |
BC044211.1 | - | 6 - 7 | 768 | 76 | |
BC091501.1 | - | 5 - 6 | 652 | 76 | |
BT006866.1 | - | 6 - 7 | 722 | 76 | |
BX647711.1 | - | 6 - 7 | 849 | 76 | |
BX648847.1 | - | 4 - 5 | 330 | 76 | |
EU668344.1 | - | 6 - 7 | 732 | 76 | |
KU178333.1 | - | 6 - 7 | 722 | 76 | |
KU178334.1 | - | 5 - 6 | 358 | 76 | |
L11932.1 | - | 5 - 6 | 633 | 76 |