The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002332.2 | NP_002323.2 | 77 - 78 | 12510 | 64 |
XM_017019303.1 | XP_016874792.1 | 78 - 79 | 12659 | 64 | |
GenBank mRNA | AB209498.1 | - | 55 - 56 | 8255 | 64 |
X13916.1 | - | 77 - 78 | 12510 | 64 |