The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316935.1 | NP_001303864.1 | 4 - 5 | 363 | 87 |
NM_016126.3 | NP_057210.2 | 4 - 5 | 560 | 87 | |
NR_133633.1 | 5 - 6 | 658 | 87 | ||
GenBank mRNA | AF100747.1 | - | 4 - 5 | 332 | 87 |
AF416725.1 | - | 3 - 4 | 287 | 87 | |
AF417114.1 | - | 3 - 4 | 287 | 87 | |
BC005245.1 | - | 4 - 5 | 352 | 87 | |
HY179788.1 | - | 4 - 5 | 364 | 87 | |
L32869.1 | - | 2 - 3 | 135 | 87 | |
X79302.1 | - | 2 - 3 | 135 | 87 |