The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318241.1 | NP_001305170.1 | 10 - 11 | 1379 | 98 |
NM_001318242.1 | NP_001305171.1 | 10 - 11 | 1443 | 98 | |
NM_001318243.1 | NP_001305172.1 | 10 - 11 | 1373 | 98 | |
NM_152710.3 | NP_689923.3 | 10 - 11 | 1376 | 98 | |
NR_134531.1 | 9 - 10 | 1294 | 98 | ||
NR_134532.1 | 10 - 11 | 1302 | 98 | ||
NR_134533.1 | 6 - 7 | 912 | 98 | ||
NR_134534.1 | 5 - 6 | 836 | 98 | ||
XM_017015853.1 | XP_016871342.1 | 11 - 12 | 1454 | 98 | |
GenBank mRNA | AK057382.1 | - | 10 - 11 | 1358 | 98 |
BC137543.1 | - | 10 - 11 | 1410 | 98 | |
BC137544.1 | - | 10 - 11 | 1344 | 98 | |
BC139728.1 | - | 9 - 10 | 1243 | 98 | |
BC144692.1 | - | 10 - 11 | 1341 | 98 | |
BC144694.1 | - | 10 - 11 | 1347 | 98 | |
HY289269.1 | - | 1 - 2 | 98 |