The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005502.3 | NP_005493.2 | 2 - 3 | 477 | 63 |
XM_005251773.2 | XP_005251830.1 | 2 - 3 | 386 | 63 | |
GenBank mRNA | AB055982.1 | - | 1 - 2 | 73 | 63 |
AB445477.1 | - | 1 - 2 | 73 | 63 | |
AF258627.1 | - | 2 - 3 | 468 | 63 | |
AK024328.1 | - | 2 - 3 | 386 | 63 | |
BC146856.1 | - | 2 - 3 | 287 | 63 | |
BX955241.1 | - | 2 - 3 | 477 | 63 |