Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CSN1S2BP
- Entrez Gene ID
-
100337616
- Gene Name
- casein alpha s2-like B, pseudogene
- Gene Aliases
-
CSN1S2B
- Chromosome Location
-
Chr.4: 70133604 - 70146704 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_033311.1 |
|
7 - 8 |
|
137 |
GenBank mRNA |
AW104440.1 |
- |
1 - 2 |
95 |
137 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
casein alpha s2-like A, pseudogene |
CSN1S2AP |
CSN1S2A |
-57.9kb
|
Sense |
Chr. - 70067386 - 70085273 on Build GRCh38 |
Downstream |
proline rich 27 |
PRR27 |
C4orf40 |
7.5kb
|
Sense |
Chr. - 70154187 - 70166609 on Build GRCh38 |
View Full Product Details