The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021138.3 | NP_066961.2 | 9 - 10 | 1195 | 56 |
XM_011518973.2 | XP_011517275.1 | 10 - 11 | 1499 | 56 | |
XM_011518974.2 | XP_011517276.1 | 9 - 10 | 2100 | 56 | |
XM_011518975.2 | XP_011517277.1 | 10 - 11 | 1427 | 56 | |
XM_011518976.2 | XP_011517278.1 | 10 - 11 | 1468 | 56 | |
XM_011518977.2 | XP_011517279.1 | 9 - 10 | 1188 | 56 | |
XM_011518978.2 | XP_011517280.1 | 9 - 10 | 2609 | 56 | |
XM_017015094.1 | XP_016870583.1 | 9 - 10 | 1243 | 56 | |
XM_017015095.1 | XP_016870584.1 | 9 - 10 | 1953 | 56 | |
GenBank mRNA | AK054686.1 | - | 8 - 9 | 1020 | 56 |
AK289722.1 | - | 10 - 11 | 1311 | 56 | |
AK294607.1 | - | 9 - 10 | 1054 | 56 | |
AK298370.1 | - | 9 - 10 | 1159 | 56 | |
BC032410.1 | - | 8 - 9 | 1181 | 56 | |
BC033810.1 | - | 9 - 10 | 1183 | 56 | |
BC043492.1 | - | 9 - 10 | 1195 | 56 | |
BC064662.1 | - | 8 - 9 | 1190 | 56 | |
BX538160.1 | - | 8 - 9 | 1137 | 56 |