The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004170.5 | NP_004161.4 | 9 - 10 | 1256 | 63 |
XM_011518007.1 | XP_011516309.1 | 10 - 11 | 1276 | 63 | |
XM_011518008.2 | XP_011516310.1 | 10 - 11 | 8521 | 63 | |
XM_011518009.2 | XP_011516311.1 | 9 - 10 | 8451 | 63 | |
XM_011518010.1 | XP_011516312.1 | 8 - 9 | 1066 | 63 | |
XM_017015042.1 | XP_016870531.1 | 9 - 10 | 1168 | 63 | |
XM_017015043.1 | XP_016870532.1 | 8 - 9 | 1109 | 63 | |
GenBank mRNA | AB008536.1 | - | 9 - 10 | 1194 | 63 |
AF037982.1 | - | 2 - 3 | 202 | 63 | |
AK313013.1 | - | 9 - 10 | 1157 | 63 | |
BC033040.1 | - | 9 - 10 | 1242 | 63 | |
U03506.1 | - | 9 - 10 | 1021 | 63 | |
U06469.1 | - | 9 - 10 | 1042 | 63 | |
U08989.1 | - | 9 - 10 | 1168 | 63 |