Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOP2B
Entrez Gene ID
7155
Gene Name
topoisomerase (DNA) II beta
Gene Aliases
TOPIIB, top2beta
Chromosome Location
Chr.3: 25597905 - 25664936 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001068.3 NP_001059.2 11 - 12 1465 131
XM_005265427.3 XP_005265484.1 11 - 12 2044 131
XM_011534057.2 XP_011532359.1 11 - 12 1454 131
GenBank mRNA AB208879.1 - 8 - 9 1024 131
AK296510.1 - 12 - 13 1520 131
AK307450.1 - 13 - 14 1465 131
M27504.1 - 7 - 8 961 131
X68060.1 - 11 - 12 1390 131

View Full Product Details