The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001068.3 | NP_001059.2 | 16 - 17 | 2072 | 103 |
XM_005265427.3 | XP_005265484.1 | 16 - 17 | 2651 | 103 | |
XM_011534057.2 | XP_011532359.1 | 16 - 17 | 2061 | 103 | |
GenBank mRNA | AB208879.1 | - | 13 - 14 | 1630 | 103 |
AK296510.1 | - | 17 - 18 | 2127 | 103 | |
AK307450.1 | - | 18 - 19 | 2072 | 103 | |
M27504.1 | - | 12 - 13 | 1568 | 103 | |
X68060.1 | - | 16 - 17 | 1997 | 103 | |
Z15115.1 | - | 2 - 3 | 227 | 103 |