The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001068.3 | NP_001059.2 | 27 - 28 | 3655 | 78 |
XM_005265427.3 | XP_005265484.1 | 27 - 28 | 4234 | 78 | |
XM_011534057.2 | XP_011532359.1 | 27 - 28 | 3644 | 78 | |
GenBank mRNA | AB208879.1 | - | 24 - 25 | 3213 | 78 |
AK296510.1 | - | 28 - 29 | 3710 | 78 | |
AK297171.1 | - | 3 - 4 | 452 | 78 | |
AK316492.1 | - | 3 - 4 | 405 | 78 | |
BC021863.1 | - | 2 - 3 | 174 | 78 | |
X53662.1 | - | 4 - 5 | 469 | 78 | |
X68060.1 | - | 27 - 28 | 3580 | 78 | |
Z15115.1 | - | 13 - 14 | 1810 | 78 |