The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317902.1 | NP_001304831.1 | 2 - 3 | 662 | 159 |
NM_024831.7 | NP_079107.6 | 3 - 4 | 727 | 159 | |
XM_005251328.2 | XP_005251385.1 | 2 - 3 | 442 | 159 | |
XM_006716485.2 | XP_006716548.1 | 3 - 4 | 513 | 159 | |
XM_017014004.1 | XP_016869493.1 | 3 - 4 | 513 | 159 | |
GenBank mRNA | AF286340.1 | - | 3 - 4 | 817 | 159 |
AF432215.1 | - | 3 - 4 | 475 | 159 | |
AK314693.1 | - | 3 - 4 | 501 | 159 | |
AY028423.1 | - | 3 - 4 | 511 | 159 | |
BX648548.1 | - | 2 - 3 | 425 | 159 |