The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317902.1 | NP_001304831.1 | 4 - 5 | 1608 | 73 |
NM_024831.7 | NP_079107.6 | 5 - 6 | 1673 | 73 | |
XM_005251328.2 | XP_005251385.1 | 4 - 5 | 1388 | 73 | |
XM_006716485.2 | XP_006716548.1 | 5 - 6 | 1459 | 73 | |
XM_017014004.1 | XP_016869493.1 | 5 - 6 | 1459 | 73 | |
GenBank mRNA | AF286340.1 | - | 5 - 6 | 1763 | 73 |
AF432215.1 | - | 5 - 6 | 1421 | 73 | |
AK055178.1 | - | 2 - 3 | 547 | 73 | |
AK314693.1 | - | 5 - 6 | 1447 | 73 | |
AY028423.1 | - | 5 - 6 | 1457 | 73 | |
AY534911.1 | - | 2 - 3 | 463 | 73 | |
BX648548.1 | - | 4 - 5 | 1371 | 73 |