Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CACNA1E
Entrez Gene ID
777
Gene Name
calcium voltage-gated channel subunit alpha1 E
Gene Aliases
BII, CACH6, CACNL1A6, Cav2.3, gm139
Chromosome Location
Chr.1: 181317712 - 181808084 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000721.3 NP_000712.2 14 - 15 2080 66
NM_001205293.1 NP_001192222.1 14 - 15 2080 66
NM_001205294.1 NP_001192223.1 14 - 15 2080 66
XM_017002243.1 XP_016857732.1 16 - 17 2745 66
XM_017002244.1 XP_016857733.1 16 - 17 2746 66
XM_017002245.1 XP_016857734.1 16 - 17 2746 66
XM_017002246.1 XP_016857735.1 16 - 17 2746 66
XM_017002247.1 XP_016857736.1 16 - 17 2745 66
XM_017002248.1 XP_016857737.1 16 - 17 2746 66
XM_017002249.1 XP_016857738.1 16 - 17 2745 66
XM_017002250.1 XP_016857739.1 16 - 17 2745 66
XM_017002251.1 XP_016857740.1 16 - 17 2403 66
GenBank mRNA AB209499.1 - 6 - 7 682 66
L27745.1 - 14 - 15 1885 66
L29384.1 - 14 - 15 2050 66
L29385.1 - 14 - 15 2050 66

View Full Product Details