Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNA1E
Entrez Gene ID
777
Gene Name
calcium voltage-gated channel subunit alpha1 E
Gene Aliases
BII, CACH6, CACNL1A6, Cav2.3, gm139
Chromosome Location
Chr.1: 181317712 - 181808084 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000721.3 NP_000712.2 31 - 32 4690 67
NM_001205293.1 NP_001192222.1 31 - 32 4690 67
NM_001205294.1 NP_001192223.1 30 - 31 4633 67
XM_017002243.1 XP_016857732.1 33 - 34 5355 67
XM_017002244.1 XP_016857733.1 33 - 34 5356 67
XM_017002245.1 XP_016857734.1 32 - 33 5344 67
XM_017002246.1 XP_016857735.1 32 - 33 5344 67
XM_017002247.1 XP_016857736.1 32 - 33 5298 67
XM_017002248.1 XP_016857737.1 33 - 34 5356 67
XM_017002249.1 XP_016857738.1 33 - 34 5355 67
XM_017002250.1 XP_016857739.1 32 - 33 5298 67
XM_017002251.1 XP_016857740.1 33 - 34 5013 67
GenBank mRNA AB209499.1 - 23 - 24 3292 67
L27745.1 - 31 - 32 4492 67
L29384.1 - 30 - 31 4603 67
L29385.1 - 31 - 32 4660 67

View Full Product Details