The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020318.2 | NP_064714.2 | 1 - 2 | 245 | 94 |
NM_021936.2 | NP_068755.2 | 1 - 2 | 245 | 94 | |
XM_011509857.1 | XP_011508159.1 | 2 - 3 | 162 | 94 | |
GenBank mRNA | AF342989.1 | - | 1 - 2 | 245 | 94 |
AF342990.1 | - | 1 - 2 | 245 | 94 | |
BC062665.1 | - | 1 - 2 | 249 | 94 | |
BC104644.1 | - | 1 - 2 | 247 | 94 | |
BC152552.1 | - | 1 - 2 | 233 | 94 |