The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008710.2 | NP_001008710.1 | 2 - 3 | 740 | 77 |
NM_001008711.2 | NP_001008711.1 | 2 - 3 | 740 | 77 | |
NM_001008712.2 | NP_001008712.1 | 2 - 3 | 740 | 77 | |
NM_006867.3 | NP_006858.1 | 2 - 3 | 740 | 77 | |
XM_017012980.1 | XP_016868469.1 | 2 - 3 | 735 | 77 | |
XM_017012981.1 | XP_016868470.1 | 2 - 3 | 735 | 77 | |
XM_017012982.1 | XP_016868471.1 | 2 - 3 | 735 | 77 | |
XM_017012983.1 | XP_016868472.1 | 2 - 3 | 1721 | 77 | |
XM_017012984.1 | XP_016868473.1 | 2 - 3 | 735 | 77 | |
XM_017012986.1 | XP_016868475.1 | 2 - 3 | 735 | 77 | |
XM_017012987.1 | XP_016868476.1 | 2 - 3 | 735 | 77 | |
XM_017012988.1 | XP_016868477.1 | 2 - 3 | 735 | 77 | |
XM_017012989.1 | XP_016868478.1 | 2 - 3 | 735 | 77 | |
XM_017012990.1 | XP_016868479.1 | 2 - 3 | 735 | 77 | |
XM_017012992.1 | XP_016868481.1 | 2 - 3 | 735 | 77 | |
GenBank mRNA | AK304858.1 | - | 3 - 4 | 558 | 77 |
AK310533.1 | - | 2 - 3 | 168 | 77 | |
BC003608.2 | - | 2 - 3 | 681 | 77 | |
BC063494.1 | - | 2 - 3 | 147 | 77 | |
BC092476.1 | - | 2 - 3 | 228 | 77 | |
D84107.1 | - | 2 - 3 | 714 | 77 | |
D84108.1 | - | 2 - 3 | 714 | 77 | |
D84109.1 | - | 2 - 3 | 714 | 77 | |
D84110.1 | - | 2 - 3 | 714 | 77 |