The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008710.2 | NP_001008710.1 | 8 - 9 | 1292 | 106 |
NM_001008711.2 | NP_001008711.1 | 9 - 10 | 1378 | 106 | |
NM_006867.3 | NP_006858.1 | 9 - 10 | 1498 | 106 | |
GenBank mRNA | AK098281.1 | - | 1 - 2 | 2309 | 106 |
BC063494.1 | - | 8 - 9 | 644 | 106 | |
CX781149.1 | - | 1 - 2 | 84 | 106 |