The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278201.1 | NP_001265130.1 | 8 - 9 | 1039 | 100 |
NM_001278206.1 | NP_001265135.1 | 8 - 9 | 1082 | 100 | |
NM_014096.3 | NP_054815.2 | 8 - 9 | 1121 | 100 | |
NM_017611.2 | NP_060081.1 | 8 - 9 | 957 | 100 | |
NM_199329.2 | NP_955361.1 | 8 - 9 | 1006 | 100 | |
GenBank mRNA | AB028927.1 | - | 8 - 9 | 992 | 100 |
AK075552.1 | - | 8 - 9 | 979 | 100 | |
AK290391.1 | - | 7 - 8 | 1019 | 100 | |
AK298028.1 | - | 8 - 9 | 1055 | 100 | |
AK300977.1 | - | 6 - 7 | 981 | 100 | |
AK311394.1 | - | 8 - 9 | 1115 | 100 | |
AK315839.1 | - | 8 - 9 | 1012 | 100 | |
AL157431.1 | - | 8 - 9 | 1026 | 100 | |
BC003163.1 | - | 8 - 9 | 958 | 100 | |
CR457391.1 | - | 6 - 7 | 674 | 100 |