The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278201.1 | NP_001265130.1 | 13 - 14 | 1738 | 84 |
NM_001278206.1 | NP_001265135.1 | 13 - 14 | 1781 | 84 | |
NM_014096.3 | NP_054815.2 | 13 - 14 | 1820 | 84 | |
NM_017611.2 | NP_060081.1 | 13 - 14 | 1656 | 84 | |
NM_199329.2 | NP_955361.1 | 13 - 14 | 1705 | 84 | |
GenBank mRNA | AB028927.1 | - | 13 - 14 | 1691 | 84 |
AK290391.1 | - | 12 - 13 | 1718 | 84 | |
AK298028.1 | - | 13 - 14 | 1754 | 84 | |
AK315839.1 | - | 13 - 14 | 1711 | 84 | |
AL157431.1 | - | 13 - 14 | 1725 | 84 | |
BC003163.1 | - | 13 - 14 | 1657 | 84 | |
CR457391.1 | - | 11 - 12 | 1373 | 84 |