Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAPPC9
Entrez Gene ID
83696
Gene Name
trafficking protein particle complex 9
Gene Aliases
IBP, IKBKBBP, MRT13, NIBP, T1, TRS120
Chromosome Location
Chr.8: 139727725 - 140458579 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160372.3 NP_001153844.1 16 - 17 2561 101
NM_001321646.1 NP_001308575.1 15 - 16 2534 101
NM_031466.7 NP_113654.4 16 - 17 2736 101
XM_011517326.2 XP_011515628.1 15 - 16 2709 101
XM_011517328.2 XP_011515630.1 16 - 17 2736 101
XM_011517330.2 XP_011515632.1 8 - 9 1101 101
XM_017013893.1 XP_016869382.1 16 - 17 2736 101
XM_017013894.1 XP_016869383.1 9 - 10 1437 101
GenBank mRNA AB067469.1 - 16 - 17 2511 101
AK026394.1 - 16 - 17 2553 101
AK027689.1 - 5 - 6 661 101
AK128755.1 - 16 - 17 2737 101
AK308815.1 - 5 - 6 832 101
AL833973.1 - 14 - 15 1836 101
AY190606.1 - 14 - 15 1815 101
BC003650.2 - 2 - 3 280 101
BC006206.2 - 14 - 15 1957 101

View Full Product Details