Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITFG1
Entrez Gene ID
81533
Gene Name
integrin alpha FG-GAP repeat containing 1
Gene Aliases
2310047C21Rik, CDA08, LNKN-1, TIP
Chromosome Location
Chr.16: 47154387 - 47461274 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030790.4 NP_110417.2 16 - 17 1891 124
GenBank mRNA AF212247.1 - 16 - 17 1799 124
AF503339.1 - 16 - 17 1681 124
AK027596.1 - 16 - 17 1682 124
AK295228.1 - 8 - 9 827 124
AK299641.1 - 11 - 12 1125 124
AL833705.1 - 10 - 11 997 124
BC006321.2 - 16 - 17 1664 124
BC024162.1 - 16 - 17 1725 124

View Full Product Details