The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001197104.1 | NP_001184033.1 | 16 - 17 | 5200 | 98 |
NM_005933.3 | NP_005924.2 | 16 - 17 | 5191 | 98 | |
XM_006718839.3 | XP_006718902.2 | 16 - 17 | 2675 | 98 | |
XM_011542829.2 | XP_011541131.1 | 17 - 18 | 5291 | 98 | |
XM_011542830.2 | XP_011541132.1 | 17 - 18 | 5288 | 98 | |
XM_011542831.2 | XP_011541133.1 | 17 - 18 | 5282 | 98 | |
XM_011542833.2 | XP_011541135.1 | 17 - 18 | 2774 | 98 | |
GenBank mRNA | AB209508.1 | - | 14 - 15 | 2735 | 98 |
D14540.1 | - | 16 - 17 | 5191 | 98 | |
L01986.1 | - | 10 - 11 | 1222 | 98 | |
L04284.1 | - | 16 - 17 | 5168 | 98 | |
L04731.1 | - | 16 - 17 | 4991 | 98 |