Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFXL1
Entrez Gene ID
152518
Gene Name
nuclear transcription factor, X-box binding like 1
Gene Aliases
CDZFP, HOZFP, OZFP, URCC5
Chromosome Location
Chr.4: 47847233 - 47914667 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278623.1 NP_001265552.1 21 - 22 2557 69
NM_001278624.1 NP_001265553.1 21 - 22 2695 69
NM_152995.5 NP_694540.3 21 - 22 2597 69
NR_103795.1 22 - 23 2737 69
GenBank mRNA AB085695.1 - 21 - 22 2553 69
AB181916.1 - 21 - 22 2677 69
AK131285.1 - 19 - 20 2143 69
AK302440.1 - 21 - 22 2555 69
AY134856.1 - 5 - 6 601 69
BC018019.1 - 4 - 5 292 69
BC051193.1 - 22 - 23 2708 69

View Full Product Details