The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278623.1 | NP_001265552.1 | 8 - 9 | 1141 | 73 |
NM_001278624.1 | NP_001265553.1 | 8 - 9 | 1279 | 73 | |
NM_152995.5 | NP_694540.3 | 8 - 9 | 1181 | 73 | |
NR_103795.1 | 8 - 9 | 1141 | 73 | ||
GenBank mRNA | AB085695.1 | - | 8 - 9 | 1137 | 73 |
AB181916.1 | - | 8 - 9 | 1261 | 73 | |
AK131285.1 | - | 6 - 7 | 727 | 73 | |
AK302440.1 | - | 8 - 9 | 1139 | 73 | |
BC051193.1 | - | 8 - 9 | 1112 | 73 |