Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAPD7
Entrez Gene ID
11044
Gene Name
PAP associated domain containing 7
Gene Aliases
LAK-1, LAK1, POLK, POLS, TRF4, TRF4-1, TRF41, TUTASE5
Chromosome Location
Chr.5: 6713952 - 6757048 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171805.1 NP_001165276.1 9 - 10 1067 88
NM_001171806.1 NP_001165277.1 3 - 4 669 88
NM_006999.4 NP_008930.1 9 - 10 1067 88
XM_005248234.3 XP_005248291.1 9 - 10 1720 88
XM_011513945.1 XP_011512247.1 7 - 8 843 88
XM_017008988.1 XP_016864477.1 9 - 10 2341 88
XM_017008989.1 XP_016864478.1 9 - 10 979 88
XM_017008990.1 XP_016864479.1 4 - 5 1525 88
GenBank mRNA AB005754.3 - 8 - 9 975 88
AF089896.1 - 9 - 10 1059 88
AK289857.1 - 8 - 9 1069 88
AK303516.1 - 3 - 4 625 88
BC084567.1 - 9 - 10 1000 88
BC117137.1 - 8 - 9 985 88
BC126106.1 - 8 - 9 985 88
BC143882.1 - 8 - 9 985 88
KC424495.1 - 9 - 10 1628 88

View Full Product Details