Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TLR4
Entrez Gene ID
7099
Gene Name
toll like receptor 4
Gene Aliases
ARMD10, CD284, TLR-4, TOLL
Chromosome Location
Chr.9: 117704175 - 117717491 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003266.3 NP_003257.1 2 - 3 513 121
GenBank mRNA AK226155.1 - 2 - 3 268 121
AK290053.1 - 2 - 3 496 121
AL710219.1 - 2 - 3 363 121
BC143841.1 - 2 - 3 235 121
U88880.1 - 2 - 3 259 121


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA652115.1 - 2 - 3 345 Chr. 9 - 117704344 - 117708710 [+] on Build GRCh38 121
GenBankmRNA DA659856.1 - 2 - 3 300 Chr. 9 - 117704388 - 117712498 [+] on Build GRCh38 121
GenBankmRNA DB078565.1 - 2 - 3 286 Chr. 9 - 117704402 - 117712507 [+] on Build GRCh38 121
GenBankmRNA DA655734.1 - 2 - 3 379 Chr. 9 - 117704311 - 117708731 [+] on Build GRCh38 121
GenBankmRNA DA651112.1 - 2 - 3 421 Chr. 9 - 117704267 - 117708723 [+] on Build GRCh38 121
GenBankmRNA DB069557.1 - 2 - 3 285 Chr. 9 - 117704403 - 117712485 [+] on Build GRCh38 121
GenBankmRNA DA326784.1 - 2 - 3 241 Chr. 9 - 117704447 - 117712555 [+] on Build GRCh38 121
GenBankmRNA AL713614.1 - 1 - 2 125 Chr. 9 - 117708197 - 117708979 [+] on Build GRCh38 121
GenBankmRNA DA990452.1 - 2 - 3 315 Chr. 9 - 117704373 - 117712441 [+] on Build GRCh38 121
GenBankmRNA DB148398.1 - 2 - 3 462 Chr. 9 - 117704226 - 117708668 [+] on Build GRCh38 121
GenBankmRNA DA338581.1 - 2 - 3 496 Chr. 9 - 117704192 - 117708664 [+] on Build GRCh38 121


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC101928797 LOC101928797 -51.2kb Sense Chr. - 117648606 - 117657027 on Build GRCh38
Downstream BMP/retinoic acid inducible neural specific 1 BRINP1 DBC1, DBCCR1, FAM5A 1.5mb Anti-sense Chr. - 119166630 - 119369461 on Build GRCh38

View Full Product Details