The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017617.4 | NP_060087.3 | 21 - 22 | 3506 | 67 |
XM_011518717.2 | XP_011517019.2 | 18 - 19 | 2798 | 67 | |
GenBank mRNA | AB209873.1 | - | 13 - 14 | 2043 | 67 |
AF308602.1 | - | 21 - 22 | 3509 | 67 | |
M73980.1 | - | 21 - 22 | 3509 | 67 |