Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOTCH1
Entrez Gene ID
4851
Gene Name
notch 1
Gene Aliases
AOS5, AOVD1, TAN1, hN1
Chromosome Location
Chr.9: 136494433 - 136545786 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017617.4 NP_060087.3 30 - 31 5641 64
XM_011518717.2 XP_011517019.2 27 - 28 4933 64
GenBank mRNA AB209873.1 - 22 - 23 4178 64
AF308602.1 - 30 - 31 5644 64
M73980.1 - 30 - 31 5644 64

View Full Product Details