The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017617.4 | NP_060087.3 | 6 - 7 | 1098 | 77 |
XM_011518717.2 | XP_011517019.2 | 3 - 4 | 390 | 77 | |
GenBank mRNA | CN431067.1 | - | 2 - 3 | 341 | 77 |