The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206696.1 | NP_001193625.1 | 4 - 5 | 717 | 92 |
NM_006147.3 | NP_006138.1 | 6 - 7 | 966 | 92 | |
GenBank mRNA | AF027292.1 | - | 5 - 6 | 730 | 92 |
AK296960.1 | - | 4 - 5 | 662 | 92 | |
AK312857.1 | - | 6 - 7 | 866 | 92 | |
BC014852.1 | - | 6 - 7 | 835 | 92 |